Learn More
B. Braun Omnifix™ Solo Luer Lock Cone Syringe
Click to view available options
Volume (Metric):
30 mL
Unit Size:
Pack of 100
Description
- Made of polypropylene
- Highly transparent barrel
- High contrast black graduation, permanent marking, ideal readability
- Safe plunger backstop
- Easy aspiration up to maximum volume
- Plunger stopper with double sealing ring for slow aspiration and injection
- Made of natural latex-free synthetic material
- Minimal residual volume
- Latex-free/PVC-free
- EN ISO 7886-1
- Single sterile-packed (box of 100 pcs)
Specifications
Specifications
| Certifications/Compliance | EN ISO 7886-1 |
| Material | Polypropylene |
| Color | Transparent |
| Description | Black graduations |
| Latex Free | Yes |
| Product Type | Luer Lock Cone |
| Disposable | Single-use |
| Quantity | 100/Box |
| Volume (Metric) | 30 mL |
| Connection Type | Luer Lock |
| Show More |