Learn More
B Braun™ Omnifix™ Solo 3-Piece Syringes
Click to view available options
Volume (Metric):
3 mL
5 mL
10 mL
20 mL
50 mL
Unit Size:
Each
Pack of 100
Description
- High visibility; transparent barrel with permanent bold graduations
- Secure backstop to prevent accidental plunger withdrawal
- Double seal stopper prevents medication leakage
- Large, stable finger grips
- Components do not contain DEHP, PVC, or other rubber tree latex
- Disposable
- Piston with double sealing ring provides slow aspiration and injection
Specifications
Specifications
| Certifications/Compliance | EN ISO 7886-1, ISO 10993 resp. DIN EN 30993 |
| Material | Polyisoprene Rubber (Tip), Polypropylene (Barrel), Polypropylene (Plunger) |
| Color | Natural |
| Description | High contrast black graduation |
| For Use With (Application) | For slow aspiration and injection |
| Latex Free | Yes |
| Product Type | Centric Luer-lock 3-part Syringe |
| Disposable | Single-use |
| Graduations | 1 mL |
| Quantity | 100/Pk. |
| Show More |