Learn More
Diba Omnifit™ BenchMark™ Replacement Frits for Chromatography Column Assemblies
Product Code. 15875544
Encompass_Preferred
Click to view available options
Porosity:
5 μm
10 μm
25 μm
Diameter (Metric) Inner:
15 mm
35 mm
50 mm
Material:
PTFE
Polyethylene
Unit Size:
Pack of 20
Product Code. 15875544
Brand: Diba 006FR5025PE
This item requires a price update prior to being ordered.
Request a Price for this item here.
Select from a variety of porosities for your media and chemistry
Replacement frits are available in PTFE or PE and in a range of pore sizes, enabling you to select the most appropriate bed supports for your application. Match the column ID to select the correct frit
Specifications
For Use With (Equipment) Chromatography Column Assemblies
Material Polyethylene
Porosity 25 μm
Size Universal
Diameter (Metric) Inner 50 mm
Diameter (English) Inner 1.9 in.
Type Replacement Chromatography Frit