Learn More
Diba Omnifit™ BenchMark™ Replacement Frits for Chromatography Column Assemblies
Click to view available options
Porosity:
5 μm
10 μm
25 μm
Diameter (Metric) Inner:
15 mm
35 mm
50 mm
Material:
PTFE
Polyethylene
Unit Size:
Pack of 20
Description
Replacement frits are available in PTFE or PE and in a range of pore sizes, enabling you to select the most appropriate bed supports for your application. Match the column ID to select the correct frit
Specifications
Specifications
| For Use With (Equipment) | Chromatography Column Assemblies |
| Material | PTFE |
| Porosity | 10 μm |
| Size | Universal |
| Diameter (Metric) Inner | 50 mm |
| Diameter (English) Inner | 1.9 in. |
| Type | Replacement Chromatography Frit |