missing translation for 'onlineSavingsMsg'
Learn More

Omni International Omni Tip™ Probes

Clear to ensure that sample is not lost in the probe

Brand:  Omni International 30750

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 11407276

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Clear to ensure that sample is not lost in the probe

Disposable to eliminate cross-contamination. May also be reused and autoclaved up to seven times.

  • Slots, instead of windows, prevent sample from being trapped
  • Probes fit tubes from 1.5 to 50mL
  • Patented

  • Soft tissue probes utilize flexible tines for gentler processing
  • Hard tissue probes have saw teeth for more aggressive processing

Specifications

Specifications

Omni Tip Soft Tissue Probe
0.27 in.
4.33 in.
Plastic
Omni TH, THQ, GLH, PDH and Omni prep 1.5 to 50mL tube homogenizer
0.25 to 30 mL
0.7 cm
11 cm
Ideal for processing liquids, soft tissues and samples that require gentle processing
25/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers