Learn More
EVOS™ Olympus 1.25X Objective, apochromat, LWD
Description
- Has long working distance (LWD) objective that is optimized for imaging of slides, cell culture dishes and flasks, and microtiter plates
- Compatible with EVOS™ Cell Imaging systems
- Offers optical performance from visible light to near infrared light
- Meets needs across spectrum of magnifications and optical specifications
Specifications
Specifications
| Type | Objective Lens |
| Product Line | Olympus |
| For Use With (Equipment) | EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Includes | Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates |
| Focus Distance | 5.11 mm |
| Lens Type | Apochromat |
| Magnification | 1.25X |
| Numerical Aperture | 0.04 |
| Quantity | 1 each |