Learn More
Olsalazine Disodium, MedChemExpress
Product Code. 16470798
Encompass_Preferred
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
Product Code. 16470798
Brand: Medchem Express HYB0174
MedChemExpress Olsalazine Disodium is an anti-inflammatory drug used in the treatment of Inflammatory Bowel Disease and Ulcerative Colitis.
Olsalazine Disodium is an anti-inflammatory drug used in the treatment of Inflammatory Bowel Disease and Ulcerative Colitis.
Specifications
Chemical Name or Material Olsalazine Disodium
CAS 6054-98-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H8N2Na2O6
Quantity 5 g
Solubility Information H2O : 50 mg/mL (144.43 mM; Need ultrasonic) ∣DMSO : 20 mg/mL (57.77 mM; Need ultrasonic)
SMILES O=C(O[Na])C1=C(O)C=CC(/N=N/C2=CC(C(O[Na])=O)=C(O)C=C2)=C1
Molecular Weight (g/mol) 346.2
Formula Weight 346.2
Percent Purity 99.36%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation