Learn More
Oligodendrocyte Marker O4 APC-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB1326A
Product Code. 18104251
  • 654.00 EUR / 100 tests
Description
Oligodendrocyte Marker O4 Monoclonal specifically detects Oligodendrocyte Marker O4 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Oligodendrocyte Marker O4
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Bovine brain corpus callosum white matter
100 Tests
Primary
Human, Mouse, Rat, Chicken
IgM
Flow Cytometry
O4
Flow Cytometry 10 uL/10^6 cells
Mouse
Affinity Purified
RUO
Detects human, mouse, rat, and chicken Oligodendrocyte Marker O4.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions