missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oligodendrocyte Marker O4 Alexa Fluor™ 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB1326T-100
This item is not returnable.
View return policy
Description
Oligodendrocyte Marker O4 Monoclonal specifically detects Oligodendrocyte Marker O4 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry.
Specifications
| Oligodendrocyte Marker O4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 5 uL/10^6 cells | |
| Mouse | |
| IgM-specific Affinity-purified from hybridoma culture supernatant | |
| RUO | |
| Detects human, mouse, rat, and chicken Oligodendrocyte Marker O4. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| O4 | |
| Alexa Fluor 594 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Bovine brain corpus callosum white matter | |
| 100 Tests | |
| Primary | |
| Human, Mouse, Rat, Chicken | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction