Learn More
Olcegepant, MedChemExpress
Product Code. 16470046
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470046 2 mg 2mg
16425957 5 mg 5mg
16409776 50 mg 50mg
16413228 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16470046 Supplier Medchem Express Supplier No. HY10095
MedChemExpress Olcegepant (BIBN-4096) is a potent and selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor with IC50 of 0.03 nM and Ki of 14.4 pM for human CGRP.
Olcegepant (BIBN-4096) is a potent and selective non-peptide antagonist of the calcitonin gene-related peptide 1 (CGRP1) receptor with IC50 of 0.03 nM and Ki of 14.4 pM for human CGRP.
Specifications
Chemical Name or Material Olcegepant
CAS 204697-65-4
Color White
Purity Grade Notes Research
Molecular Formula C38H47Br2N9O5
Quantity 2 mg
Synonym BIBN-4096 BIBN 4096BS
Solubility Information DMSO : ≥ 50 mg/mL (57.49 mM) ∣1M HCl : 50 mg/mL (57.49 mM; ultrasonic and adjust pH to 1 with HCl)
SMILES O=C1N(CC2=C(N1)C=CC=C2)C3CCN(C(N[C@@H](C(N[C@H](C(N4CCN(C5=CC=NC=C5)CC4)=O)CCCCN)=O)CC6=CC(Br)=C(C(Br)=C6)O)=O)CC3
Molecular Weight (g/mol) 869.65
Formula Weight 869.65
Percent Purity 97.92%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation