Learn More
Kern Sohn OIML Class M3 Block Test Weight
Click to view available options
Weight (Metric):
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
50 kg
Shape:
Cylindrical
Knob
Rectangular
Unit Size:
Each
Description
- Lacquered cast iron
Specifications
Specifications
| Classification | M3 |
| Material | Cast Iron |
| Weight (Metric) | 50 kg |
| Shape | Rectangular |