Learn More
Kern Sohn OIML Class M3 Block Test Weight
Product Code. 17392393
Encompass_Preferred
Click to view available options
Weight (Metric):
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
50 kg
Shape:
Cylindrical
Knob
Rectangular
Unit Size:
Each
Product Code. 17392393
Brand: Kern Sohn 36689
Only null left
Add to basket
Add to basket
Single test weight constructed of cast iron.
  • Lacquered cast iron
Specifications
Classification M3
Material Cast Iron
Weight (Metric) 50 kg
Shape Rectangular