Learn More
Sartorius™ OIML Class E2 Proof Line Weight Set with DAkkS Certificate
Product Code. 11333364 Shop All Sartorius Products
Click to view available options
Mass:
1 g to 1 kg
1 g to 200 g
1 mg to 500 mg
Density:
2,950 kg/m3 (1 to 5mg), 8,600 kg/m3 (10mg to 500mg)
7,950 kg/m3
Material:
Aluminum (1 to 5 mg), Nickel Silver (10 to 500 mg)
Special Stainless Steel
Unit Size:
Each
Product Code. 11333364
Brand: Sartorius™ YCS011252AC02
Only null left
Add to basket
Add to basket
Includes the same features and properties as the individual weights of this series, along with the corresponding maximum permissible errors. Sartorius™ OIML Class E2 Weight Set with DAkkS Certificate is supplied with gloves, tweezers and brush in high-quality plastic case
Values 1 to 5mg:
Aluminum leaf weights with density of 2,650 kg/m3
Values of 10 to 500mg:
Nickel silver construction with density of 8,600 kg/m3.
Values of 1g and higher:
Polished, special stainless steel knob weights with density of 7,950kg/m3
  • Highly corrosion resistant
  • Non-magnetizable
  • Tolerances comply with OIML R111-1
  • Includes sturdy plastic carry case
  • DAkkS Certified
TRUSTED_SUSTAINABILITY
Specifications
Product Type Proof Line Weight Set
Certifications/Compliance DAkkS calibration
Classification OIML Class E2
Material Aluminum (1 to 5 mg), Nickel Silver (10 to 500 mg)
Density 2,950 kg/m3 (1 to 5mg), 8,600 kg/m3 (10mg to 500mg)
Mass 1 mg to 500 mg