Learn More
Kern Sohn OIML Class E2 Compact Test Weight
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
Each
Description
- Compact shape
- Polished stainless steel
Specifications
Specifications
| Classification | E2 |
| Material | Stainless Steel |
| Weight (Metric) | 5 g |
| Tolerance | ± 0.05 mg |