Learn More
Kern Sohn OIML Class E2 Compact Test Weight
Product Code. 17302003
Encompass_Preferred
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
Each
Product Code. 17302003
Brand: Kern Sohn 31601
Only null left
Add to basket
Add to basket
Single test weight constructed of stainless steel.
  • Compact shape
  • Polished stainless steel
Specifications
Classification E2
Material Stainless Steel
Weight (Metric) 1 g
Tolerance ± 0.03 mg