Learn More
asecos™ Oil Sorbents Mat
Product Code. 17836516 Shop All asecos Products
Change view
Click to view available options
Weight (Metric):
7 kg
8 kg
Unit Size:
Pack of 100
Pack of 200
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) unitSize
17836516 8 kg Pack of 100
17866346 7 kg Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17836516 Supplier asecos™ Supplier No. 9934
The Oil Sorbents Mat is water repellant for the safe absorption of oils, petrol, diesel, etc.
  • Very strong durability
  • Suitable for heavy and long-lasting use
  • Tightly woven fibres, high abrasion resistance and low-lint surface
  • Outstanding absorption performance
  • Large absorption capacity (up to 16 time of own weight)
  • Light for smaller leakages and prevention
  • Heavy for larger leakages or absorption of puddles
Specifications
Color Blue
Material Heavy-duty Cloth
Type Oil Only Sorbent Pad
Length (English) 15.75 in.
Length (Metric) 400.00 mm
Width (English) 15.75 in.
Width (Metric) 400.00 mm
Weight (English) 17.64 lb.
Weight (Metric) 8 kg