missing translation for 'onlineSavingsMsg'
Learn More

OICR-9429, MedChemExpress

Product Code. 16423267
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16423267

Brand: Medchem Express HY16993

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress OICR-9429 is high affinity WD repeat domain 5 (WDR5) inhibitor, competitively blocks WDR5 interaction with MLL protein via binding the central peptide-binding pocket of WDR5. OICR-9429 can suppress histone H3K4 trimethylation and can be used for the research of various cancers including non-MLL-rearranged leukaemia, colon, pancreatic, prostate cancer and bladder cancer (BCa) .

OICR-9429 is high affinity WD repeat domain 5 (WDR5) inhibitor, competitively blocks WDR5 interaction with MLL protein via binding the central peptide-binding pocket of WDR5. OICR-9429 can suppress histone H3K4 trimethylation and can be used for the research of various cancers including non-MLL-rearranged leukaemia, colon, pancreatic, prostate cancer and bladder cancer (BCa) .

Specifications

Chemical Name or Material OICR-9429
Physical Form Solid
CAS 1801787-56-3
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C29H32F3N5O3
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 32 mg/mL (57.60 mM)
SMILES O=C(C1=CNC(C=C1C(F)(F)F)=O)NC2=CC(C3=CC(CN4CCOCC4)=CC=C3)=CC=C2N5CCN(C)CC5
Molecular Weight (g/mol) 555.59
Formula Weight 555.59
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.