missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OCT6 Monoclonal Antibody (KT110), Invitrogen™

Product Code. p-200088893
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30197901 200 μL 200µg
30203733 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30197901 Supplier Invitrogen™ Supplier No. MA555998

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

POU3F1 is a member of the POU domain family of proteins, and regulates events during neurogenesis and myelination.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCT6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone KT110
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene POU3F1
Gene Accession No. P20267, P21952
Gene Alias 6-Oct; LOW QUALITY PROTEIN: POU domain, class 3, transcription factor 1; Oct6; Oct-6; octamer binding factor oct6; octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; Otf6; OTF-6; POU class 3 homeobox 1; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1; Scip; suppressed cAMP-inducible POU; Test1; Testes-1; Tst1; Tst-1
Gene Symbols POU3F1
Host Species Mouse
Immunogen Recombinant mouse Oct6 (proprietary range information).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18991, 192110
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.