missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAS2 Monoclonal Antibody (OTI4B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802771S
This item is not returnable.
View return policy
Description
OAS2 Monoclonal antibody specifically detects OAS2 in Human samples. It is validated for Western BlotSpecifications
| OAS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P29728 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4939 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4B11 | |
| Unconjugated | |
| OAS2 | |
| (2-5')oligo(A) synthase 2; (2'-5')oligo(A) synthetase 2; 2',5'-oligoadenylate synthetase-like 11; 2,5-oligoadenylate synthetase-like 11; 2'-5' oligoadenylate synthetase 2; 2'-5' oligoadenylate synthetase-like 11; 2-5A synthase 2; 2'-5'-oligoadenylate synthase 2; 2'-5'-oligoadenylate synthetase 2; 2'-5'-oligoadenylate synthetase 2, 69/71kDa; A synthetase 2; MGC78578; Oas2; Oasl11; p69 OAS / p71 OAS; p69OAS / p71OAS | |
| Human recombinant protein fragment corresponding to amino acids 60-335 of human OAS2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction