missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14955-1-AP-20UL
This item is not returnable.
View return policy
Description
OAS1 is an interferon inducible protein that may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. It binds double-stranded RNA and polymerizes ATP into PPP(A2′P5′A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. This protein is associated with different subcellular fractions such as mitochondrial, nuclear, and rough/smooth microsomal fractions.Specifications
| OAS1 | |
| Polyclonal | |
| Unconjugated | |
| OAS1 | |
| (2 5)oligo(A) synthase 1, 2 5 oligoadenylate synthase 1, 2 5A synthase 1, E18/E16, IFI 4, OAS1, OIAS, OIASI, p46/p42 OAS | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 192281, 246730, 4938 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00973, P11928, Q05961 | |
| OAS1, Oas1a | |
| OAS1 Fusion Protein Ag6793 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction