Learn More
Conductivity Calibration Pouches, Oakton™
Product Code. 12393780 Shop All Cole Parmer Products
Change view
Click to view available options
Conductivity:
702 ppm NaCl, 1000 ppm 442, 745 ppm KCl
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Conductivity unitSize
12393780 702 ppm NaCl, 1000 ppm 442, 745 ppm KCl Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12393780 Supplier Cole Parmer Supplier No. 3565311
Calibrate on the go with these single-use 20mL calibration pouches. Oakton™ Conductivity Calibration Pouches are sealed to ensure freshness.
  • Convenient calibration pouches are portable
  • A μS-to-ppm conversion table is printed on each pack for quick reference
  • Accuracy is ±1% at 25±C
  • Calibration pouches include a NIST-traceable calibration report supplied by the manufacture
Specifications
Chemical Name or Material Cole-Parmer Conductivity Standards
Packaging Bottle
Conductivity 702 ppm NaCl, 1000 ppm 442, 745 ppm KCl
Technique Calibration of Conductivity Instrument
Color Clear
pH 4.7 to 7.8
Boiling Point 100°C (212°F)
For Use With (Equipment) Conductivity Meter
Solubility Soluble
Solution Type Calibration Solution
Type Conductivity Standard
Quantity 20 Single-use Pouches