missing translation for 'onlineSavingsMsg'
Learn More

o-Toluic acid, 98+%

Product Code. p-201086 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
1kg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10563111 5 g Glass Bottle 5g
10224810 100 g Plastic bottle 100g
10336880 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10563111 Supplier Thermo Scientific Acros Supplier No. 139040050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-90-1 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-90-1
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00002477
InChI Key ZWLPBLYKEWSWPD-UHFFFAOYSA-N
Synonym o-toluic acid, o-toluylic acid, orthotoluic acid, 2-toluic acid, toluic acid, benzoic acid, 2-methyl, o-methylbenzoic acid, 2-methyl-benzoic acid, benzoic acid, methyl, o-toluate
PubChem CID 8373
ChEBI CHEBI:36632
IUPAC Name 2-methylbenzoic acid
SMILES CC1=CC=CC=C1C(=O)O

Specifications

Melting Point 103°C to 105°C
Color White to Yellow
Density 1.06
Boiling Point 258°C to 259°C
Flash Point 148°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass Bottle
Linear Formula CH3C6H4CO2H
Quantity 5 g
Beilstein 09, 462
Merck Index 14, 9535
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and chloroform
Formula Weight 136.15
Percent Purity 98+%
Physical Form Needle-like Crystalline Solid
Chemical Name or Material o-Toluic acid, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • o-Toluic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.