Learn More
DWK Life Sciences O-Ring, red, FEP coated, not suitable for desiccators
Shop All DWK Life Sciences Products
Click to view available options
Diameter (Metric) Outside:
75 mm
110 mm
133 mm
157 mm
215 mm
Unit Size:
Each
Description
Flat flange vessels are large-volume reaction vessels consisting of a lower part and a lid. These two parts are held together by a clamping ring. The red O-ring made of silicone is seamlessly coated with FEP and serves as an optimal seal for flat flange vessels with an annular groove. It prevents jamming of the ground surfaces between the base and the lid and is convincing due to its good elasticity in combination with outstanding chemical resistance.
- Accessories for flat flange vessels, comprising an elastic, silicone core with seamless FEP coating that encloses the ring
- The combination of these high-quality materials achieves good elasticity in conjunction with outstanding chemical resistance
Specifications
Specifications
| For Use With (Application) | Tight closure of flange vessels |
| Quantity | 1 Ea. |
| Material | Silicone, FEP |
| Diameter (Metric) Outside | 110 mm |