missing translation for 'onlineSavingsMsg'
Learn More

O,O'-Butane-1,4-diyl bis(4-hydroxybutyl) diterephthalate, TRC

Product Code. 30331512
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331512 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30331512 Supplier Toronto Research Chemicals Supplier No. TRCB57449010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-O-[4-[4-(4-hydroxybutoxycarbonyl)benzoyl]oxybutyl] 1-O-(4-hydroxybutyl) benzene-1,4-dicarboxylate
CAS 29278-67-9
Molecular Formula C28 H34 O10
Quantity 10 mg
Synonym O,O'-Butane-1,4-diyl bis(4-hydroxybutyl) diterephthalate, 1,4-Benzenedicarboxylic acid, 1,4-butanediyl bis(4-hydroxybutyl) ester (9CI), Terephthalic acid, tetramethylene bis(4-hydroxybutyl) ester (8CI), 1,4-Butanediol, terephthalate (3:2) (8CI)
InChI Formula InChI=1S/C28H34O10/c29-15-1-3-17-35-25(31)21-7-11-23(12-8-21)27(33)37-19-5-6-20-38-28(34)24-13-9-22(10-14-24)26(32)36-18-4-2-16-30/h7-14,29-30H,1-6,15-20H2
SMILES OCCCCOC(=O)c1ccc(cc1)C(=O)OCCCCOC(=O)c2ccc(cc2)C(=O)OCCCCO
IUPAC Name 4-O-[4-[4-(4-hydroxybutoxycarbonyl)benzoyl]oxybutyl] 1-O-(4-hydroxybutyl) benzene-1,4-dicarboxylate
Molecular Weight (g/mol) 530.564
Formula Weight 530.215
Product Title
Select an issue

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.