missing translation for 'onlineSavingsMsg'
Learn More

O,O-Bis(4-chlorophenyl) Phosphorochloridothioate, TRC

Product Code. 30333119
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30333119 Supplier Toronto Research Chemicals Supplier No. TRCB5850151G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O,O-Bis(4-chlorophenyl) Phosphorochloridothioate
CAS 55526-70-0
Molecular Formula C12H8Cl3O2PS
Quantity 1 g
Synonym O,O-Bis(p-chlorophenyl) Chlorothiophosphate
InChI Formula InChI=1S/C12H8Cl3O2PS/c13-9-1-5-11(6-2-9)16-18(15,19)17-12-7-3-10(14)4-8-12/h1-8H
SMILES Clc1ccc(OP(=S)(Cl)Oc2ccc(Cl)cc2)cc1
IUPAC Name chloro-bis(4-chlorophenoxy)-sulfanylidene-λ^{5}-phosphane
Molecular Weight (g/mol) 353.59
Formula Weight 351.9048
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.