missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Desmethyl O-TBDMS Primaquine, TRC
Specifications
Specifications
| Chemical Name or Material | O-Desmethyl O-TBDMS Primaquine |
| Analyte or Component Names | O-Desmethyl O-TBDMS Primaquine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H33 N3 O Si |
| Synonym | N4-(6-((tert-Butyldimethylsilyl)oxy)quinolin-8-yl)pentane-1,4-diamine |
| InChI Formula | InChI=1S/C20H33N3OSi/c1-15(9-7-11-21)23-18-14-17(24-25(5,6)20(2,3)4)13-16-10-8-12-22-19(16)18/h8,10,12-15,23H,7,9,11,21H2,1-6H3 |
| Solution Type | Neat |
| SMILES | CC(CCCN)Nc1cc(O[Si](C)(C)C(C)(C)C)cc2cccnc12 |
| IUPAC Name | 4-N-[6-[tert-butyl(dimethyl)silyl]oxyquinolin-8-yl]pentane-1,4-diamine |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?