missing translation for 'onlineSavingsMsg'
Learn More

Melford NZCYM Broth

Product Code. p-200087174
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 30018609

Brand: Melford N720001000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Enriched medium for cultivating recombinant strains of E.

Enriched medium for cultivating recombinant strains of E.Coli and propagating theta bacteriophage.

Dissolve 22 grams in 1 liter distilled water. Sterilize by autoclaving for 15 minutes.

Casein Digest Protein: 10 g/L

Yeast Extract: 5 g/L

Casamino Acids: 1 g/L

Magnesium Sulfate: 0.98 g/L

Sodium Chloride: 5 g/L

Specifications

Quantity 1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.