missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford NZCYM Broth
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
Description
Enriched medium for cultivating recombinant strains of E.
Enriched medium for cultivating recombinant strains of E.Coli and propagating theta bacteriophage.
Dissolve 22 grams in 1 liter distilled water. Sterilize by autoclaving for 15 minutes.
Casein Digest Protein: 10 g/L
Yeast Extract: 5 g/L
Casamino Acids: 1 g/L
Magnesium Sulfate: 0.98 g/L
Sodium Chloride: 5 g/L
Specifications
Specifications
| Quantity | 250 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction