Learn More
Masterflex™ Nylon Male Pipe Adapter
Designed for quick and easy assembly without tools
Brand:  Masterflex™ 30621-47
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11714959
  • 23.60 EUR / Pack of 10
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
  • Adapts from a NPT male thread to a hose barb
  • Barbed fittings are designed for quick and easy assembly without tools
  • Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30psi)
  • Available in packs of ten
Specifications
Specifications
Male Pipe Adapter
Not Autoclavable
1/4 in.
1/4 in. NPT(M)
Ethylene Oxide
Translucent
Barb x NPT(M) Connection
Nylon
1/4 in.
1/4 in.
1/4 in. NPT(M)