missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: LESDMKRKGK LKNKGSKRKK EDLQEVDGEI EAVLQKKAKA RGLEFQISNV KFEDVGGNDM TLKEVCKMLI HMRHPEVYHH LG Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 78%.
Gasdermin D is a member of the gasdermin family. Members of this family appear to play a role in regulation of epithelial proliferation. Gasdermin D has been suggested to act as a tumor suppressor. Alternatively spliced transcript variants have been described.
Specifications
Specifications
| Antigen | NVL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NVL |
| Gene Accession No. | O15381, Q9DBY8 |
| Gene Alias | 1200009I24Rik; Nuclear valosin-containing protein-like; nuclear VCP-like; nuclear VCP-like protein; Nvl; NVL2; NVLp |
| Gene Symbols | NVL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
