Learn More
Thermo Scientific™ Nunc™ Cell Culture Treated Flasks with Solid Caps
BIOPHARMA
Product Code. p-4524973
Click to view available options
Culture Area:
175 cm²
25 cm²
80 cm²
Closure Type:
Solid
Vent/Close
Unit Size:
Pack of 160
Pack of 32
Pack of 50
Product Code. 10068780
Brand: Thermo Scientific™ 153732
Edge
Achieve maximum adhesion for a broad range of cell types.
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific Nunc Cell Culture Treated Flasks with Solid Caps. These flasks receive the proprietary Nunclon Delta surface treatment to ensure consistent cell growth. The vent/close caps with Y marks allow easy visual verification of venting position while promoting sufficient gas exchange.
Features
  • Nunclon Delta Certification, including testing on four different cell types, provided in each case
  • Short wide necks, straight side walls, and 1/3-turn caps optimized for robotic operation
  • Vent/close caps with 'Y' marks. Any of the legs in the 'Y' pointing up indicates cap in venting position.
  • Sterilized by gamma irradiation and certified to be non-pyrogenic
Warranty: 90 days
TRUSTED_SUSTAINABILITY
Specifications
Description Nunc Flask 80cm², Solid Cap, Straight Neck, 50/Cs
Material Polystyrene
Neck Style Straight
No. per Case 50
No. per Pack 5
Sterility Sterile
Closure Type Solid
Culture Area 80 cm²
Surface Treatment Nunclon Delta
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.