missing translation for 'onlineSavingsMsg'
Learn More

Numidargistat dihydrochloride, MedChemExpress

Product Code. p-200100781
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16483668

Brand: Medchem Express HY101979A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Numidargistat (CB-1158) dihydrochloride is a potent and orally active inhibitor of arginase, with IC50s of 86 nM and 296 nM for recombinant human arginase 1 and recombinant human arginase 2, respectively. Immuno-oncology agent.

Numidargistat (CB-1158) dihydrochloride is a potent and orally active inhibitor of arginase, with IC50s of 86 nM and 296 nM for recombinant human arginase 1 and recombinant human arginase 2, respectively. Immuno-oncology agent.

Specifications

Chemical Name or Material Numidargistat dihydrochloride
Color White
Purity Grade Notes Research
Molecular Formula C11H24BCl2N3O5
Quantity 10 mg
Synonym CB-1158 dihydrochlorideINCB01158 dihydrochloride
Solubility Information DMSO : 55 mg/mL (152.76 mM; Need ultrasonic) ∣H2O : 37.78 mg/mL (104.93 mM; Need ultrasonic)
SMILES O=C([C@@H](N)C)N1C[C@H](CCCB(O)O)[C@](N)(C(O)=O)C1.Cl.Cl
Molecular Weight (g/mol) 360.04
Formula Weight 360.04
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.