Learn More
Macherey-Nagel™ NucleoSpin™ Blood XL, Maxi kit for DNA from blood
Description
NucleoSpin™ Blood XL is designed for the rapid, large-scale preparation of highly pure genomic DNA from up to 10 mL whole blood, serum, plasma or other body fluids. Buffy coat volumes up to 2mL can be processed easily. The binding capacity of the NucleoSpin™ Blood XL columns is about 700μg of genomic DNA. The kit allows preparation of pure genomic DNA from 2mL to 10 mL of human whole blood with an A260/280 ratio between 1.7 and 1.9.Typical DNA yields – depending on the concentration of leukocytes in the respective blood sample – are in the range of 20μg to 30μg per mL of healthy human blood. Blood treated either with EDTA, citrate, heparin or CPDA can be used. Time for preparation of genomic DNA from blood with NucleoSpin™ Blood XL is approximately 1 hour. The obtained DNA can be used directly as template for PCR* reactions or can be subjected to endonuclease digestion. Kit components: NucleoSpin™ Blood XL columns, 50 mL collecting tubes, 50 mL elution tubes, buffers, proteinase K.
- Silica membrane technology
- Yield: 200μg to 300μg
- Sample material: ≤10 mL/108 cells
- Elution volume: 500μL to 2, 000μL
- Binding capacity: >1,000μg
- Preparation time: 1hr/prep
- Format: mini spin column
- No use of organic solvents For rapid isolation of genomic DNA from whole blood, serum, plasma or other body fluids.
Specifications
Specifications
| Detection Method | Silica-membrane Technology |
| Format | Maxi Spin Column |
| For Use With (Application) | Whole Blood, Treated with Citrate, EDTA, Heparin, CPDA |
| Includes | 2mL Collection Tubes, Buffers, Proteinase K |
| No. of Reactions | 10 |
| Product Type | NucleoSpin™ Blood XL Column |
| Quantity | 1 Each |
| Test Time | 60min. |
| Yield | 200 to 300μg |
Safety and Handling
Hazard Category
- Skin sensitiser Category 1