Learn More
Macherey-Nagel™ NucleoSpin™ 8 Tissue, 8-well kit for DNA from cells and tissue
Description
NucleoSpin™ 8 Tissue is designed for the rapid simultaneous preparation of highly pure genomic DNA from animal and human tissue, e.g. mouse and rat tails.
From a 1.2cm mouse tail tip section (age of mice 4 to 6 weeks) up to 30μg to 40μg of pure genomic DNA can be prepared (typical yields 15μg to 25μg). The obtained DNA can be used directly for PCR*, Southern blotting or any kind of enzymatic reaction.Parallel lysis of large sample numbers can be performed in standard tubes or in 96 well blocks alternatively. NucleoSpin™ Tissue binding strips can be used under vacuum or centrifugation in a centrifuge with deep buckets allowing processing of NucleoSpin™ 8 well strips stacked on 96 well blocks. The method is based on a specially treated silica membrane with a high binding capacity for nucleic acids. The obtained DNA can be used directly for PCR*, solution blotting, or any kind of enzymatic reaction.Kit components: NucleoSpin™ Tissue binding strips, buffers, proteinase K, MN wash plates, MN square-well blocks, racks with tube strips, self-adhering foil. For further details on this product please contact Customer Services.- Silica membrane technology
- Format: 8 well strip
- Preparation time: 1hr (after lysis)
- Sample material: <20mg tissue or 106 cultured cells, bacteria
- Yield: 15μg to 25μg
- Elution volume: 100μL to 200μL
- DNA binding capacity: 40μg
- Fragment size: 300bp to 30kb
- No use of organic solvents
Specifications
Specifications
| Format | 8-well Strip |
| Isolation Technology | Silica Membrane Technology |
| For Use With (Application) | For Manual and Automated Isolation of DNA |
| No. of Reactions | 60 x 8 |
| Product Type | NucleoSpin™ 8 Tissue Binding Strip |
| Sample Volume | <20 mg human/animal tissue, <106 cells |
| Test Time | 20min. |
| Yield | 15 to 25μg |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : guanidine hydrochloride,2-propanol