Learn More
Macherey-Nagel™ Nucleosil™ 120-5 C18 EC HPLC Separation Column
Product Code. 10411852 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric):
4 mm
4.6 mm
Unit Size:
Each
Product Code. 10411852
Brand: Macherey-Nagel™ 720051.46
Only null left
Add to basket
Add to basket
Column Format: Standard Column; Carbon Load: 11%; Pore Size: 120 Å; Column Phase: Reversed Phase
Specifications
Column Format Standard Column
Diameter (Metric) 4.6 mm
Length (Metric) 125 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 120 Å
Surface Area 200 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 11%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Type EC Separative Column