Learn More
Macherey-Nagel™ Nucleodur™ Amino (NH2) EC Mixed Mode HPLC Column, 5 μm, 4 mm x 125 mm
Product Code. 12418487 Shop All Macherey-Nagel Products
Click to view available options
Diameter (Metric):
4 mm
Unit Size:
Each
Product Code. 12418487
Brand: Macherey-Nagel™ 760720.40
Only null left
Add to basket
Add to basket
Column Format: Analytical Column; Carbon Load: 2.50%; Pore Size: 110 Å; Column Phase: Reversed Phase, Normal Phase, Ion Exchange, HILIC
Specifications
Column Format Analytical Column
Crosslinking Aminopropyl
Diameter (Metric) 4 mm
For Use With HPLC
Length (Metric) 125 mm
Max. Pressure 8700 psi (600 bar)
Mobile Phase Compatibility n-heptane
Particle Size 5 μm
pH 2 to 8
Pore Size 110 Å
Quantity 1 Pk.
Solvents n-heptane
Surface Area 340 m²/g
Temperature 60°C
USP Type L8
Stationary Phase Amino (NH2)
Packing Material Silica
Carbon Load 2.5%
Column Type Mixed Mode
Endcapped No
Phase Reversed Phase, Normal Phase, Ion Exchange, HILIC
Material Silica
Type EC Column
Technique EC