missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NTCP Polyclonal Antibody
GREENER_CHOICE

Product Code. 15565242
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15565242 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15565242 Supplier Invitrogen™ Supplier No. PA542101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: VFSLAMKGDM NLSIVMTTCS TFCALGMMPL LLYIYSRGIY DGDLKDKVPY Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 93%; Rat: 100%.

Sodium/bile acid cotransporters are integral membrane glycoproteins that participate in the enterohepatic circulation of bile acids. Two homologous transporters are involved in the reabsorption of bile acids, one absorbing from the intestinal lumen, the bile duct, and the kidney with an apical localization (SLC10A2), and the other being found in the basolateral membranes of hepatocytes (SLC10A1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NTCP
Applications Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC10A1
Gene Accession No. Q14973
Gene Alias bile acid cotransporting polypeptide; cell growth-inhibiting gene 29 protein; GIG29; growth-inhibiting protein 29; hepatic sodium-dependent bile acid transporter; LOW QUALITY PROTEIN: sodium/bile acid cotransporter; MGC128766 protein; Na(+)/bile acid cotransporter; Na(+)/taurocholate transport protein; Na/taurocholate cotransporting polypeptide; NA-dependent cholate transporting protein; Ntcp; Ntcp1; SBACT; Slc10a1; sodium bile acid cotransporting polypeptide; sodium/bile acid cotransporter; sodium/tauro; sodium/taurocholate cotransporter; sodium/taurocholate cotransporting polypeptide; sodium-dependent bile acid cotransporter; sodium-dependent taurocholate cotransporting polypeptide; sodium-taurocholate cotransporting polypeptide; solute carrier family 10 (sodium/bile acid cotransporter family), member 1; solute carrier family 10 (sodium/bile acid cotransporter), member 1; solute carrier family 10 member 1; solute carrier family 10, member 1
Gene Symbols SLC10A1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SLC10A1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6554
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.