missing translation for 'onlineSavingsMsg'
Learn More

NSC5844, MedChemExpress

Product Code. 16472758
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16472758

Brand: Medchem Express HY100033

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC5844 (RE-640) is a 4-aminoquinoline derivative, with antitumor and antimalarial activity.

NSC5844 (RE-640) is a 4-aminoquinoline derivative, with antitumor and antimalarial activity.

Specifications

Chemical Name or Material NSC5844
Physical Form Solid
CAS 140926-75-6
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H16Cl2N4
For Use With (Application) Cancer-programmed cell death
Synonym RE-640
Solubility Information DMSO : 1 mg/mL (2.61 mM; ultrasonic and warming and heat to 60°C)
SMILES ClC1=CC=C2C(NCCNC3=CC=NC4=CC(Cl)=CC=C34)=CC=NC2=C1
Molecular Weight (g/mol) 383.27
Formula Weight 383.27
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.