missing translation for 'onlineSavingsMsg'
Learn More

NSC 74859, MedChemExpress

Product Code. 16436328
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436328

Brand: Medchem Express HY15146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC 74859 (S3I-201) is a selective Stat3 inhibitor with an IC50 of 86 μM.

NSC 74859 (S3I-201) is a selective Stat3 inhibitor with an IC50 of 86 μM.

Specifications

Chemical Name or Material NSC 74859
CAS 501919-59-1
Color Gray
Purity Grade Notes Research
Molecular Formula C16H15NO7S
Quantity 10 mg
Synonym S3I-201
Solubility Information DMSO : 100 mg/mL (273.70 mM; Need ultrasonic)
SMILES OC1=CC(NC(COS(=O)(C2=CC=C(C=C2)C)=O)=O)=CC=C1C(O)=O
Molecular Weight (g/mol) 365.36
Formula Weight 365.36
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.