missing translation for 'onlineSavingsMsg'
Learn More

NSC 601980 (analog), MedChemExpress

Product Code. p-200099338
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488796

Brand: Medchem Express HYB1714

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NSC 601980 analog is the analog of the NSC601980, NSC601980 shows antitumor activity in the yeast screening experiment, which can inhibit cell proliferation in the COLO 205 and HT29 with Log GI 50 of -6.6 and -6.9 respectively.

NSC 601980 analog is the analog of the NSC601980, NSC601980 shows antitumor activity in the yeast screening experiment, which can inhibit cell proliferation in the COLO 205 and HT29 with Log GI 50 of -6.6 and -6.9 respectively.

Specifications

Chemical Name or Material NSC 601980 (analog)
CAS 91757-46-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H14N4
Quantity 100 mg
Solubility Information DMSO : ≥ 30 mg/mL (119.86 mM)
SMILES CC1=NC2=CC=CC=C2N/C1=N/NC3=CC=CC=C3
Molecular Weight (g/mol) 250.3
Formula Weight 250.3
Percent Purity 96.43%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.