missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NRBF2 Monoclonal Antibody (K40009_14D6), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA556173

 View more versions of this product

Product Code. 30198099

  • 846.00 EUR / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence of this protein is as follows: GQSPLSQKYS PSTEKCLPEI QGIFDRDPDT LLYLLQQKSE PAEPCIGSKA PKDDKTIIEE QATKIADLKR HVEFLVAENE RLRKENKQLK AEKARLLKGP IEKELDVDAD

NRBF2 (nuclear receptor-binding factor 2) modulates transcriptional activation by target nuclear receptors. It is involved in starvation-induced autophagy probably by its association with PI3K complex I (PI3KC3-C1). NRBF2 stabilizes the PI3KC3-C1 assembly and enhances ATG14-linked lipid kinase activity of PIK3C3.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NRBF2
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
Q96F24
Nrbf2
Recombinant human NRBF2 (immunogen range is 111-220).
200 μg
Primary
Human
Antibody
IgG2b
Immunoprecipitation, Western Blot
K40009_14D6
Unconjugated
Nrbf2
Comodulator of PPAR and RXR; comodulator of PPAR and RXR 1; comodulator of PPAR and RXR 2; COPR; COPR1; COPR2; Nrbf2; NRBF-2; nuclear receptor binding factor 2; nuclear receptor binding factor-2; nuclear receptor-binding factor 2
Mouse
Protein A
RUO
29982
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.