missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPSR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552442
This item is not returnable.
View return policy
Description
Immunogen sequence: DILDNFNLLP DTQERFYASV IIQNLPALNS AINPLIYCVF SSSISFPCRE RRSQDSRMTF RERTERHEMQ ILSKP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 84%.
May be active in signaling pathway in an autocrine or paracrine fashion in several tissues. Receptor for neuropeptide S, it may mediate its action, such as inhibitory effects, on cell growth. Involved in pathogenesis of asthma and other IgE-mediated diseases.
Specifications
| NPSR1 | |
| Polyclonal | |
| Unconjugated | |
| NPSR1 | |
| 9330128H10Rik; ASRT2; G protein-coupled receptor 154; G protein-coupled receptor PGR14; Gpr154; GPRA; G-protein coupled receptor 154; G-protein coupled receptor for asthma susceptibility; G-protein coupled receptor PGR14; MVTR; Neuropeptide S receptor; neuropeptide S receptor 1; NPSR; Npsr1; Pgr14; RGD1564154; vasopressin receptor-related receptor 1; vasotocin receptor; VRR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 387129 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6W5P4 | |
| NPSR1 | |
| Recombinant protein corresponding to Human NPSR1. Recombinant protein control fragment (Product #RP-89585). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction