missing translation for 'onlineSavingsMsg'
Learn More

NPS-2143 hydrochloride, MedChemExpress

Product Code. 16473518
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473518

Brand: Medchem Express HY10171

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NPS-2143 hydrochloride (SB-262470A hydrochloride), an orally active calcilytic agent, is a selective and potent calcium ion-sensing receptor (CaSR) antagonist. NPS-2143 hydrochloride (SB-262470A hydrochloride) blocks increases in cytoplasmic Ca2+ concentrations (IC50=43 nM) elicited by activating the Ca2+ receptor in HEK 293 cells expressing the human Ca2+ receptor.

NPS-2143 hydrochloride (SB-262470A hydrochloride), an orally active calcilytic agent, is a selective and potent calcium ion-sensing receptor (CaSR) antagonist. NPS-2143 hydrochloride (SB-262470A hydrochloride) blocks increases in cytoplasmic Ca2+ concentrations (IC50=43 nM) elicited by activating the Ca2+ receptor in HEK 293 cells expressing the human Ca2+ receptor.

Specifications

Chemical Name or Material NPS-2143 hydrochloride
CAS 324523-20-8
Color White
Purity Grade Notes Research
Molecular Formula C24H26Cl2N2O2
Quantity 10 mg
Synonym SB-262470A hydrochloride
Solubility Information DMSO : ≥ 100 mg/mL (224.53 mM) ∣H2O : 1.85 mg/mL (4.15 mM; ultrasonic and warming and heat to 60°C)
SMILES N#CC1=C(OC[C@@H](CNC(C)(C)CC2=CC=C3C=CC=CC3=C2)O)C=CC=C1Cl.[H]Cl
Molecular Weight (g/mol) 445.38
Formula Weight 445.38
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.