missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ NPBM™ Neural Progenitor Basal Medium
Description
- Basal medium optimized for growth and in vitro differentiation of neural progenitor cells (NHNP)
- May be used for experimental applications including drug development, neurotoxicity, neurogenesis, electrophysiology, neurotransmitter disorders, and central nervous system function.
- Growth medium BulletKit™ (CC-3209) as well as a differentiation medium BulletKit™ (CC-3229) available.
Specifications
Specifications
| Cell Type | Neural Cells |
| Product Type | Basal Medium |
| Quantity | 200 mL |
| Color | White |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?