missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Npas4 Antibody (S408-79), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59332JF549
This item is not returnable.
View return policy
Description
Npas4 Monoclonal antibody specifically detects Npas4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Npas4 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| BHLHE79, bHLHe79neuronal PAS domain-containing protein 4, Class E basic helix-loop-helix protein 79, Le-PAS, neuronal PAS domain protein 4, Neuronal PAS4, NXFHLH-PAS transcription factor NXF, PASD10PAS domain-containing protein 10 | |
| Fusion protein amino acids 597-802 (C-terminus) of rat Npas4. | |
| 0.1 mL | |
| Cellular Signaling, Epigenetics, Neuroscience | |
| 266743 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| S408-79 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction