missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Noxa Antibody (SR3425), Novus Biologicals™
Rabbit Monoclonal Antibody
327.00 EUR - 382.00 EUR
Specifications
| Antigen | Noxa |
|---|---|
| Clone | SR3425 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041692
|
Novus Biologicals™
NBP3-27768-50ul |
50 μL |
327.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041788
|
Novus Biologicals™
NBP3-27768-100ul |
100 μL |
382.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Noxa Monoclonal antibody specifically detects Noxa in Human samples. It is validated for Western BlotSpecifications
| Noxa | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, PMA-Induced Protein 1, Protein Noxa | |
| A synthesized peptide derived from human Noxa (Uniprot #: Q13794) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3425 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cancer, Cellular Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Mitochondrial Mediated Pathway, Plasma Membrane Markers, Stem Cell Markers, Tumor Suppressors | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 5366 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title