Learn More
Novus Biologicals™ Recombinant Human HIF-1 alpha Protein
Highly purified. Generating reliable and reproducible results.
Brand: Novus Biologicals™ NBC1-18422
This item is not returnable.
View return policy
Description
An un-tagged recombinant protein corresponding to the amino acids 530-826 of Human HIF-1 alpha The Recombinant Human HIF-1 alpha Protein is derived from E. coli. The Recombinant Human HIF-1 alpha Protein has been validated for the following applications: SDS-Page, Block/Neutralize.Specifications
| Q16665 | |
| 3091 | |
| HIF-1 alpha Protein | |
| 0.1mg | |
| Hif-1a (530-826 aa) Human, Recombinant, E.coli (NP_001521). | |
| RUO | |
| Unlabeled |
| 20mM Tris-HCl buffer (pH 7.5) 1mM DTT | |
| 32.8kDa | |
| Protein | |
| E. Coli | |
| −80°C. Avoid freeze-thaw cycles. | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 | |
| Yes |