Learn More
Novobiocin Sodium Salt, TRC
Product Code. 30009797
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009797 5 g 5g
30009873 10 g 10g
30009789 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30009797 Supplier Toronto Research Chemicals Supplier No. TRCN8885005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1476-53-5
Molecular Formula C31 H35 N2 O11 . Na
Molecular Weight (g/mol) 634.61
Synonym Benzamide, N-[7-[[3-O-(aminocarbonyl)-6-deoxy-5-C-methyl-4-O-methyl-α-L-lyxo-hexopyranosyl]oxy]-4-hydroxy-8-methyl-2-oxo-2H-1-benzopyran-3-yl]-4-hydroxy-3-(3-methyl-2-buten-1-yl)-, sodium salt (1:1) (ACI), Benzamide, N-[7-[[3-O-(aminocarbonyl)-6-deoxy-5-C-methyl-4-O-methyl-α-L-lyxo-hexopyranosyl]oxy]-4-hydroxy-8-methyl-2-oxo-2H-1-benzopyran-3-yl]-4-hydroxy-3-(3-methyl-2-butenyl)-, monosodium salt (9CI), Novobiocin, monosodium salt (8CI), Novobiocin, sodium deriv. (6CI, 7CI), Albamycin, Albamycin (capsule), Albamycin sodium, Cathomycin sodium, Cathomycin sodium lyovac, Monosodium novobiocin, Novobiocin monosodium, Novobiocin sodium, Novobiocin sodium salt, NSC 2382, Sodium albamycin, Sodium novobiocin, Vulcamycin
IUPAC Name sodium;7-[(2R,3R,4S,5R)-4-carbamoyloxy-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy-3-[[4-hydroxy-3-(3-methylbut-2-enyl)benzoyl]amino]-8-methyl-2-oxochromen-4-olate
SMILES [Na+].CO[C@@H]1[C@@H](OC(=O)N)[C@@H](O)[C@H](Oc2ccc3C(=C(NC(=O)c4ccc(O)c(CC=C(C)C)c4)C(=O)Oc3c2C)[O-])OC1(C)C
Specifications
Quantity 5 g
InChI Formula InChI=1S/C31H36N2O11.Na/c1-14(2)7-8-16-13-17(9-11-19(16)34)27(37)33-21-22(35)18-10-12-20(15(3)24(18)42-28(21)38)41-29-23(36)25(43-30(32)39)26(40-6)31(4,5)44-29;/h7,9-13,23,25-26,29,34-36H,8H2,1-6H3,(H2,32,39)(H,33,37);/q;+1/p-1/t23-,25+,26-,29-;/m1./s1
Formula Weight 634.2139
Percent Purity >95%
Chemical Name or Material Novobiocin sodium
Grade HPLC