Learn More
TOKU-E™ Novobiocin ReadyMade™ Solution
Product Code. 30044018
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30044018 1 mL 1mL
30044032 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30044018 Supplier TOKU-E™ Supplier No. N0891ML
Novobiocin ReadyMade™ Solution is provided as a sterile-filtered solution of Novobiocin Sodium in water.
  • CAS Number: 1476-53-5
  • Molecular Formula: C31H35N2NaO11
  • Molecular Weight: 634.61 g/mol
  • Storage Conditions: -20°C, Protect from light
  • Form: Liquid
  • Source: Streptomyces spp.
  • Spectrum: Novobiocin Sodium targets primarily Gram-positive organisms and is commonly used to differentiate between coagulase-negative Staphylococcus species (all but S. aureus and a few others), with S. saprophyticus being resistant to novobiocin in culture
  • Mechanism of Action: Aminocoumarin antibiotics target DNA-gyrase, an enzyme which relieves strain as DNA unwinds during replication and transcription. It also inhibits mammalian topoisomerase, both are ATP-dependent enzymes involved with cell division
Specifications
Chemical Name or Material Novobiocin Sodium
Physical Form Liquid
CAS 1476-53-5
Source Streptomyces spp.
Packaging 2 mL plastic vial
Sterile Filtered Yes, 0.22 μm
Sterility Sterile
Color Yellow
Concentration 100 to 105 mg/mL
Quantity 1 mL
Purity Grade Notes >90% (HPLC)
Assay Percent Range >850 μg/mg
Assay Percent Notes Novobiocin Sodium, Dried Basis
Molecular Formula C31H35N2NaO11
Product Line ReadyMade™ Solutions
Shipping Condition Ice Pack
IUPAC Name sodium;7-[(2R,3R,4S,5R)-4-carbamoyloxy-3-hydroxy-5-methoxy-6,6-dimethyloxan-2-yl]oxy-3-[[4-hydroxy-3-(3-methylbut-2-enyl)benzoyl]amino]-8-methyl-2-oxochromen-4-olate
Molecular Weight (g/mol) 634.61 g/mol
Formula Weight 634.61 amu
Percent Purity >90%
Grade Antibiotic
Concentration or Composition (by Analyte or Components) Novobiocin Sodium ≥5 to <10%