Learn More
Invitrogen™ ZOOM™ IPG Strips pH 9-12
Description
Requires
ZOOM IPGRunner System
- First dimension separation, isoelectric focusing (IEF), is complete in less than three hours
- The mini-gel design of the system is easy to handle, eliminates mineral oil overlays, and allows processing of up to 12 samples at once for high-throughput usage
- Strips are 7cm long and contain a thin layer of polyacrylamide gel that includes a fixed pH gradient
- Each strip is clearly labeled with a unique identifying number, pH range, and orientation marks
- Strips are supplied attached to a tri-fold card for easy access and removal
2D Gel Electrophoresis, IsoElectric Focusing, Protein Gel Electrophoresis, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Dry Ice
Specifications
Specifications
| Content And Storage | The ZOOM™ IPGRunner™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid, Gel Tension Wedge, Buffer Dam, and the Mini- Cell chamber. The ZOOM™ IPGRunner™ Combo Kit includes the ZOOM™ IPGRunner™ Mini-Cell, electrode wicks, sealing tape, 10 ZOOM™ IPGRunner™ Cassettes, and 12 ZOOM™ Strips, pH 3-10 NL. The ZOOM™ IPGRunner™ Retrofit Kit for XCell SureLock™ Mini-Cell includes the ZOOM™ IPGRunner™ Buffer Core and Lid. The ZOOM™ IPGRunner™ Cassettes include electrode wicks and sealing tape. |
| Gel Size | Mini |
| pH Range | 9.0 - 12.0 |
| Quantity | 12 strips |
| Shipping Condition | Dry Ice |
| Product Line | ZOOM |
| Product Type | IPG Strip |
| For Use With (Equipment) | XCell SureLock Mini-Cell |