Learn More
Invitrogen™ Denaturing Cell Extraction Buffer
Product Code. 10561665
Change view
Click to view available options
Quantity:
100 ml
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10561665 100 ml Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10561665 Supplier Invitrogen™ Supplier No. FNN0091
Suitable for use in ELISA, Western blotting, sample/assay prep
Denaturing Cell Extraction Buffer can be used for Invitrogen phosphoELISAs (i.e., ERK, JNK) that require sample treatment or boiling. After lysing cells with Denaturing Cell Extraction Buffer, lysates will not require sample treatment or boiling.
Build Your Own Immunoassay, Cell Analysis, Cell Lysis, Immunoassays, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Order Info
Shipping condition: Dry ice
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer (-5 to -30°C).
Product Type Cell Extraction Buffer
Quantity 100 ml
Shipping Condition Dry Ice